Tisk…

Anti-ABCE1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Dodavatel: Bioss
BS-2948R-A750
BOSSBS-2948R-A750EA 25340 CZK
BOSSBS-2948R-A750
Anti-ABCE1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Protilátky
ABCE1 is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the OABP subfamily. It is alternatively referred to as the RNase L inhibitor as it functions to block the activity of ribonuclease L(referenced from entrez gene).

Type: Primary
Antigen: ABCE1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 331-430/599
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Objednat

Produktová specifikace

Název antigenu ABCE1
Environmentally Preferable
Synonyma antigenu 2''-5''-oligoadenylate-binding protein|RNS 4I|Ribonuclease 4 Inhibitor|ABCE1_HUMAN|2'5' oligoadenylate binding protein|ATP-binding cassette sub-family E member 1|ABC 38|HuHP 68|ATP binding cassette sub family E OABP member 1|ABCE 1|Ribonuclease L 2' 5' oligoisoadenylate synthetase dependent inhibitor|HuHP68|OABP|2' 5' oligoadenylate binding protein|ABCE1|Ribonuclease L 2'5' oligoisoadenylate synthetase dependent inhibitor|RNASELI|RLI|RNS4I|ABC38|RNase L Inhibitor|ATP binding cassette sub family E member 1|RNASEL1|Ribonuclease L inhibitor
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Alexa Fluor® 750
Reaktivita Rat
Hostitel Rabbit
Identifikační číslo genu 6059
Izotyp IgG
Western blot Yes
Imunochemie Yes
ImunoFluorescence Yes
Modifikace Unmodified
Epitop 331-430/599
Imunogen KLH conjugated synthetic peptide derived from human ABCE1
Formulář Liquid
Molekulární hmotnost 67 kDa
Čištění Purified by Protein A
Koncentrace 1 µg/µl
Skladovací pufr Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Skladovací teplota Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Transportní teplota 4 °C
Testované aplikace ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR