Tisk…

Anti-SLC16A3 Rabbit Polyclonal Antibody (Cy3®)

Dodavatel: Bioss
BS-2698R-CY3
BOSSBS-2698R-CY3EA 25630 CZK
BOSSBS-2698R-CY3
Anti-SLC16A3 Rabbit Polyclonal Antibody (Cy3®)
Protilátky
Proton-linked monocarboxylate transporter. Catalyzes the rapid transport across the plasma membrane of many monocarboxylates such as lactate, pyruvate, branched-chain oxo acids derived from leucine, valine and isoleucine, and the ketone bodies acetoacetate, beta-hydroxybutyrate and acetate (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC16A3
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR