Tisk…

Anti-RGN Rabbit Polyclonal Antibody (Cy7®)

Dodavatel: Bioss
BS-2665R-CY7
BOSSBS-2665R-CY7EA 25630 CZK
BOSSBS-2665R-CY7
Anti-RGN Rabbit Polyclonal Antibody (Cy7®)
Protilátky
Gluconolactonase with low activity towards other sugar lactones, including gulonolactone and galactonolactone. Can also hydrolyze diisopropyl phosphorofluoridate and phenylacetate (in vitro). Calcium-binding protein. Modulates Ca(2+) signaling, and Ca(2+)-dependent cellular processes and enzyme activities (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RGN
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR