Tisk…

Anti-CCL16 Rabbit Polyclonal Antibody

Dodavatel: Bioss
BS-2481R
BOSSBS-2481REA 20650 CZK
BOSSBS-2481R
Anti-CCL16 Rabbit Polyclonal Antibody
Protilátky
LEC (liver expressed cytokine) displays chemotactic activity for lymphocytes and monocytes but not for neutrophils. It also shows a potent myelosuppressive activity and suppresses proliferation of myeloid progenitor cells. Its expression is upregulated by IL10.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CCL16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR