Tisk…
Anti-ATG7 Rabbit Polyclonal Antibody (Cy5®)
Dodavatel: Bioss
BS-2432R-CY5
BOSSBS-2432R-CY5EA 25890 CZK
BOSSBS-2432R-CY5
Anti-ATG7 Rabbit Polyclonal Antibody (Cy5®)
Protilátky
E1-like activating enzyme involved in the 2 ubiquitin-like systems required for cytoplasm to vacuole transport (Cvt) and autophagy. Activates ATG12 for its conjugation with ATG5 as well as the ATG8 family proteins for their conjugation with phosphatidylethanolamine. Both systems are needed for the ATG8 association to Cvt vesicles and autophagosomes membranes. Required for autophagic death induced by caspase-8 inhibition. Required for mitophagy which contributes to regulate mitochondrial quantity and quality by eliminating the mitochondria to a basal level to fulfill cellular energyMore Product Information