Tisk…

Anti-GNMT Rabbit Polyclonal Antibody (Cy5.5®)

Dodavatel: Bioss
BS-2396R-CY5.5
BOSSBS-2396R-CY5.5EA 25630 CZK
BOSSBS-2396R-CY5.5
Anti-GNMT Rabbit Polyclonal Antibody (Cy5.5®)
Protilátky
Catalyzes the methylation of glycine by using S-adenosylmethionine (AdoMet) to form N-methylglycine (sarcosine) with the concomitant production of S-adenosylhomocysteine (AdoHcy). Possible crucial role in the regulation of tissue concentration of AdoMet and of metabolism of methionine.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GNMT
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR