Tisk…
Anti-FOXA2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Dodavatel: Bioss
BS-2358R-A350
BOSSBS-2358R-A350EA 25660 CZK
BOSSBS-2358R-A350
Anti-FOXA2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
Transcription factor that is involved in embryonic development, establishment of tissue-specific gene expression and regulation of gene expression in differentiated tissues. Is thought to act as a 'pioneer' factor opening the compacted chromatin for other proteins through interactions with nucleosomal core histones and thereby replacing linker histones at target enhancer and/or promoter sites. Binds DNA with the consensus sequence 5'-[AC]A[AT]T[AG]TT[GT][AG][CT]T[CT]-3' (By similarity). In embryonic development is required for notochord formation. Involved in the development of multipleMore Product Information
Objednat
Produktová specifikace
Symbol antigenu FOXA2
Environmentally Preferable
Název antigenu Forkhead Box A2
Synonyma antigenu Forkhead box protein A2|Hepatocyte nuclear factor 3-beta|HNF3B|HNF-3-beta|TCF3B
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Alexa Fluor® 350
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 3170
Izotyp IgG
Imunochemie Yes
Průtoková cytometrie Yes
Imunogen 200-250/457
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C