Tisk…

Anti-Pectinesterase inhibitor 18 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Dodavatel: Bioss
BS-2219R-A750
BOSSBS-2219R-A750EA 25340 CZK
BOSSBS-2219R-A750
Anti-Pectinesterase inhibitor 18 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Protilátky
Pectinesterase inhibitor 18.

Type: Primary
Antigen: Pectinesterase inhibitor 18
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 451-557/557
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Produktová specifikace

Název antigenu Pectinesterase inhibitor 18
Environmentally Preferable
Synonyma antigenu Pectin methylesterase inhibitor 18|AtPMEpcrA|PE 18|Contains:Bunctional pectinesterase 18/rRNA N-glycosylase|Contains: Pectinesterase inhibitor 18|Pectinesterase/pectinesterase inhibitor 18|AtPME4|Pectin methylesterase 18|PME18_ARATH|Pectin methylesterase 4
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Alexa Fluor® 750
Reaktivita
Hostitel Rabbit
Izotyp IgG
Western blot Yes
Imunochemie Yes
ImunoFluorescence Yes
Modifikace Unmodified
Epitop 451-557/557
Imunogen KLH conjugated synthetic peptide derived from Arabidopsis thaliana pectinesterase inhibitor 18 Pectinesterase
Formulář Liquid
Molekulární hmotnost 58 kDa
Čištění Purified by Protein A
Koncentrace 1 µg/µl
Skladovací pufr Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Skladovací teplota Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Transportní teplota 4 °C
Testované aplikace ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR