Tisk…
Anti-MKI67 Rabbit Polyclonal Antibody (Cy7®)
Dodavatel: Bioss
BS-2130R-CY7
BOSSBS-2130R-CY7EA 25660 CZK
BOSSBS-2130R-CY7
Anti-MKI67 Rabbit Polyclonal Antibody (Cy7®)
Protilátky
Proliferation Marker Ki67 antigen is the prototypic cell cycle related nuclear protein, expressed by proliferating cells in all phases of the active cell cycle (G1, S, G2 and M phase). It is absent in resting (G0) cells. Ki67 are useful in establishing the cell growing fraction in neoplasms (immunohistochemically quantified by determining the number of Ki67 positive cells among the total number of resting cells = Ki67 index). In neoplastic tissues the prognostic value is comparable to the tritiated thymidine labelling index. The correlation between low Ki67 index and histologically low gradeMore Product Information