Tisk…

Anti-CNGA2 Rabbit Polyclonal Antibody (Cy3®)

Dodavatel: Bioss
BS-2074R-CY3
BOSSBS-2074R-CY3EA 25630 CZK
BOSSBS-2074R-CY3
Anti-CNGA2 Rabbit Polyclonal Antibody (Cy3®)
Protilátky
Odorant signal transduction is probably mediated by a G-protein coupled cascade using cAMP as second messenger. The olfactory channel can be shown to be activated by cyclic nucleotides which leads to a depolarization of olfactory sensory neurons.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CNGA2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR