Tisk…

Anti-AQP9 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Bioss
BS-2060R-HRP
BOSSBS-2060R-HRPEA 25890 CZK
BOSSBS-2060R-HRP
Anti-AQP9 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Forms a channel with a broad specificity. Mediates passage of a wide variety of non-charged solutes including carbamides, polyols, purines, and pyrimidines in a phloretin- and mercury-sensitive manner, whereas amino acids, cyclic sugars, Na(+), K(+), Cl(-), and deprotonated monocarboxylates are excluded. Also permeable to urea and glycerol.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: AQP9
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Produktová specifikace

Symbol antigenu AQP9
Název antigenu Aquaporin 9
Environmentally Preferable
Synonyma antigenu HsT17287|SSC1|AQP-9
Druh protilátky Primární
Klonovalita Polyclonal
Spojení HRP (Horseradish Peroxidase)
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 366
Izotyp IgG
Western blot Yes
Imunochemie Yes
Imunogen 211-246/295
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR