Tisk…

Anti-AQP9 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Dodavatel: Bioss
BS-2060R-A555
BOSSBS-2060R-A555EA 25660 CZK
BOSSBS-2060R-A555
Anti-AQP9 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Protilátky
Forms a channel with a broad specificity. Mediates passage of a wide variety of non-charged solutes including carbamides, polyols, purines, and pyrimidines in a phloretin- and mercury-sensitive manner, whereas amino acids, cyclic sugars, Na(+), K(+), Cl(-), and deprotonated monocarboxylates are excluded. Also permeable to urea and glycerol.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: AQP9
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR