Tisk…

Anti-PCNA Rabbit Polyclonal Antibody

Dodavatel: Bioss
BS-2007R
BOSSBS-2007REA 5560 CZK
BOSSBS-2007R
Anti-PCNA Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PCNA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR