Tisk…

Anti-MTOR Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Dodavatel: Bioss
BS-1992R-A647
BOSSBS-1992R-A647EA 25660 CZK
BOSSBS-1992R-A647
Anti-MTOR Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
The protein encoded by this gene belongs to a family of phosphatidylinositol kinase-related kinases. These kinases mediate cellular responses to stresses such as DNA damage and nutrient deprivation. This protein acts as the target for the cell-cycle arrest and immunosuppressive effects of the FKBP12-rapamycin complex. The ANGPTL7 gene is located in an intron of this gene. [provided by RefSeq, Sep 2008]

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MTOR
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Produktová specifikace

Symbol antigenu MTOR
Environmentally Preferable
Název antigenu Mechanistic Target of Rapamycin
Synonyma antigenu FRAP1|mTOR|m-TOR|RAPT1|RAFT1|FKBP-Rapamycin Associated Protein|Mechanistic Target Of Rapamycin|EC 2.7.11.1|FRAP|FRAP2|Rapamycin Target Protein|FK506 binding protein 12-rapamycin associated protein|Mammalian Target Of Rapamycin|Serine/Threonine-Protein Kinase MTOR|MTOR
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Alexa Fluor® 647
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Identifikační číslo genu 2475
Izotyp IgG
Imunochemie Yes
Imunogen 2436-2492/2549
Formulář liquid
Čištění Purified by Protein A
Koncentrace 1 μg/μl
Křížová adsorbce No
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Skladovací teplota Store at 4°C for 12 months
Transportní teplota 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR