Tisk…

Anti-PEG3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Bioss
BS-1870R-HRP
BOSSBS-1870R-HRPEA 25890 CZK
BOSSBS-1870R-HRP
Anti-PEG3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Induces apoptosis in cooperation with SIAH1A. Acts as a mediator between p53/TP53 and BAX in a neuronal death pathway that is activated by DNA damage. Acts synergistically with TRAF2 and inhibits TNF induced apoptosis through activation of NF-kappa-B (By similarity). Possesses a tumor suppressing activity in glioma cells.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: PEG3
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR