Tisk…
Anti-ox-LDL Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Dodavatel: Bioss
BS-1698R-A350
BOSSBS-1698R-A350EA 24950 CZK
BOSSBS-1698R-A350
Anti-ox-LDL Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
Low-density lipoprotein (LDL) is the carrier protein for cholesterol in the blood. LDL binds to its receptor on the capillary walls and thereby mediates the uptake and clearence of cholesterol from the circulation. In atherosclerotic lesions oxidatively modified LDL is found and oxidized LDL is specifically recognized and ingested by macrophages via scavenger receptor A and CD36. Oxidized LDL may be a marker of atherosclerosis but the precise changes in oxidized LDL are not well described. Low-density lipoprotein oxidised with Cu2SO4.
Recommended Dilutions: IF(IHC-P): 1:50-200