Tisk…

Anti-DCN Rabbit Polyclonal Antibody

Dodavatel: Bioss
BS-1695R
BOSSBS-1695REA 20050 CZK
BOSSBS-1695R
Anti-DCN Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: DCN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR