Tisk…
Anti-B3GAT1/CD57 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Dodavatel: Bioss
BS-1635R-A488
BOSSBS-1635R-A488EA 25660 CZK
BOSSBS-1635R-A488
Anti-B3GAT1/CD57 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Protilátky
Involved in the biosynthesis of L2/HNK-1 carbohydrate epitope on glycoproteins. Can also play a role in glycosaminoglycan biosynthesis. Substrates include asialo-orosomucoid (ASOR), asialo-fetuin, and asialo-neural cell adhesion molecule. Requires sphingomyelin for activity: stearoyl-sphingomyelin was the most effective, followed by palmitoyl-sphingomyelin and lignoceroyl-sphingomyelin. Activity was demonstrated only for sphingomyelin with a saturated fatty acid and not for that with an unsaturated fatty acid, regardless of the length of the acyl group (By similarity).
Type: Primary