Tisk…
Anti-IGFBPL1 Rabbit Polyclonal Antibody
Dodavatel: Bioss
BS-15569R
BOSSBS-15569REA 20650 CZK
BOSSBS-15569R
Anti-IGFBPL1 Rabbit Polyclonal Antibody
Protilátky
IGFBPL1 is a secreted IGF (Insulin-like growth factor) binding protein that is known to contain an Ig-like C2-type (immunoglobulin-like) domain, an IGFBP N-terminal domain and a Kazal-like domain. IGF-binding proteins characteristically act to extend the half-life of IGFs and may influence the growth promoting effects of the IGFs. The interaction of IGFBPs with IGFs can affect cell surface receptors, specifically, IGFBPs may enhance or decrease a cells insulin sensitivity. IGFBPL1 has been found to be down-regulated in multiple tumors and thus may be a likely tumor suppressor candidate.More Product Information
Objednat
Produktová specifikace
Symbol antigenu IGFBPL1
Environmentally Preferable
Synonyma antigenu bA113O24.1|insulin-like growth factor binding protein related protein 4|IGFBPRP4|IGFBP-related protein 10|IBPL1_HUMAN|insulin-like growth factor-binding protein-like 1|IGFBP-RP4|insulin-like growth factor-binding-related protein 4.
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Izotyp IgG
Western blot Yes
Imunochemie Yes
Formulář liquid
Čištění Purified by Protein A.
Koncentrace 1 ug/ul
Skladovací pufr Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Transportní teplota 4 °C