Tisk…

Anti-T140 Rabbit Polyclonal Antibody (HRP)

Dodavatel: Bioss
BS-15559R-HRP
BOSSBS-15559R-HRPEA 25630 CZK
BOSSBS-15559R-HRP
Anti-T140 Rabbit Polyclonal Antibody (HRP)
Protilátky
IFT140 is a gene encodes one of the subunits of the intraflagellar transport (IFT) complex A. Intraflagellar transport is involved in the genesis, resorption and signaling of primary cilia. The primary cilium is a microtubule-based sensory organelle at the surface of most quiescent mammalian cells, that receives signals from its environment, such as the flow of fluid, light or odors, and transduces those signals to the nucleus. Loss of the corresponding protein in mouse results in renal cystic disease.

Type: Primary
Antigen: T140
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public ImmunogenMore Product Information
Objednat

Produktová specifikace

Název antigenu T140
Environmentally Preferable
Synonyma antigenu Intraflagellar transport 140 homolog Chlamydomonas|Gs114|140_HUMAN|WD and tetratricopeptide repeats protein 2|WDTC2
Druh protilátky Primární
Klonovalita Polyclonal
Spojení HRP (Horseradish Peroxidase)
Reaktivita Rat
Hostitel Rabbit
Izotyp IgG
ELISA Yes
Imunochemie Yes
Modifikace Unmodified
Epitop 1151-1250/1462
Imunogen KLH conjugated synthetic peptide derived from human IFT140
Formulář Liquid
Molekulární hmotnost 165 kDa
Čištění Purified by Protein A
Koncentrace 1 µg/µl
Skladovací pufr Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Skladovací teplota Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Transportní teplota 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR