Tisk…
Anti-PG-C/Pepsinogen 2 Rabbit Polyclonal Antibody (Cy3®)
Dodavatel: Bioss
BS-1551R-CY3
BOSSBS-1551R-CY3EA 25630 CZK
BOSSBS-1551R-CY3
Anti-PG-C/Pepsinogen 2 Rabbit Polyclonal Antibody (Cy3®)
Protilátky
This gene encodes an aspartic proteinase that belongs to the peptidase family A1. The encoded protein is a digestive enzyme that is produced in the stomach and constitutes a major component of the gastric mucosa. This protein is also secreted into the serum. This protein is synthesised as an inactive zymogen that includes a highly basic prosegment. This enzyme is converted into its active mature form at low pH by sequential cleavage of the prosegment that is carried out by the enzyme itself. Polymorphisms in this gene are associated with susceptibility to gastric cancers. Serum levels ofMore Product Information
Objednat
Produktová specifikace
Environmentally Preferable
Název antigenu PG-C/Pepsinogen 2
Synonyma antigenu Pepsinogen C|PGC|PGII|PEPC|Gastricsin
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Cy3®
Reaktivita Human
Hostitel Rabbit
Identifikační číslo genu 5225
Izotyp IgG
Imunochemie Yes
ImunoFluorescence Yes
Modifikace Unmodified
Epitop 21-120/388
Imunogen KLH conjugated synthetic peptide derived from human PG-C
Formulář Liquid
Molekulární hmotnost 42 kDa
Čištění Purified by Protein A
Koncentrace 1 µg/µl
Skladovací pufr Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Skladovací teplota Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Transportní teplota 4 °C
Testované aplikace ICC