Tisk…
Anti-PG-C/Pepsinogen 2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Dodavatel: Bioss
BS-1551R-A555
BOSSBS-1551R-A555EA 25430 CZK
BOSSBS-1551R-A555
Anti-PG-C/Pepsinogen 2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Protilátky
This gene encodes an aspartic proteinase that belongs to the peptidase family A1. The encoded protein is a digestive enzyme that is produced in the stomach and constitutes a major component of the gastric mucosa. This protein is also secreted into the serum. This protein is synthesised as an inactive zymogen that includes a highly basic prosegment. This enzyme is converted into its active mature form at low pH by sequential cleavage of the prosegment that is carried out by the enzyme itself. Polymorphisms in this gene are associated with susceptibility to gastric cancers. Serum levels ofMore Product Information