Tisk…

Anti-COMT Rabbit Polyclonal Antibody (Cy5)

Dodavatel: Bioss
BS-15507R-CY5
BOSSBS-15507R-CY5EA 25630 CZK
BOSSBS-15507R-CY5
Anti-COMT Rabbit Polyclonal Antibody (Cy5)
Protilátky
Catalyses the O-methylation, and thereby the inactivation, of catecholamine neurotransmitters and catechol hormones. Also shortens the biological half-lives of certain neuroactive drugs, like L-DOPA, alpha-methyl DOPA and isoproterenol.

Type: Primary
Antigen: COMT
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 121-220/271
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR