Tisk…
Anti-C3Orf34 Rabbit Polyclonal Antibody (Cy7®)
Dodavatel: Bioss
BS-15175R-CY7
BOSSBS-15175R-CY7EA 25630 CZK
BOSSBS-15175R-CY7
Anti-C3Orf34 Rabbit Polyclonal Antibody (Cy7®)
Protilátky
C3orf34 (chromosome 3 open reading frame 34), also known as MGC14126, is a 163 amino acid protein encoded by a gene that maps to human chromosome 3q29. Chromosome 3 is made up of approximately 214 million bases encoding over 1,100 genes. Notably, there is a chemokine receptor gene cluster and a variety of human cancer related loci on chromosome 3. Particular regions of the chromosome 3 short arm are deleted in many types of cancer cells. Key tumor suppressing genes on chromosome 3 encode apoptosis mediator RASSF1, cell migration regulator HYAL1 and angiogenesis suppressor SEMA3B. MarfanMore Product Information