Tisk…

Anti-C2orf71 Rabbit Polyclonal Antibody

Dodavatel: Bioss
BS-15158R
BOSSBS-15158REA 20650 CZK
BOSSBS-15158R
Anti-C2orf71 Rabbit Polyclonal Antibody
Protilátky
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: C2orf71
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR