Tisk…

Anti-FASN/Fatty Acid Synthase Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Dodavatel: Bioss
BS-1498R-A555
BOSSBS-1498R-A555EA 25660 CZK
BOSSBS-1498R-A555
Anti-FASN/Fatty Acid Synthase Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Protilátky
Fatty acid synthetase catalyses the formation of long-chain fatty acids from acetyl-CoA, malonyl-CoA and NADPH. This multifunctional protein has 7 catalytic activities and an acyl carrier protein.

Type: Primary
Antigen: FASN/Fatty Acid Synthase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 555
Public Immunogen Range: 651-750/2511
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Goat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR