Tisk…

Anti-Gastrokine 1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Bioss
BS-14441R-HRP
BOSSBS-14441R-HRPEA 25630 CZK
BOSSBS-14441R-HRP
Anti-Gastrokine 1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Has mitogenic activity and may be involved in maintaining the integrity of the gastric mucosal epithelium.

Type: Primary
Antigen: Gastrokine 1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 101-185/185
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR