Tisk…

Anti-DALRD3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Dodavatel: Bioss
BS-14192R-A750
BOSSBS-14192R-A750EA 25340 CZK
BOSSBS-14192R-A750
Anti-DALRD3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Protilátky
arginine-tRNA ligase activity

Type: Primary
Antigen: DALRD3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 352-450/543
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR