Tisk…

Anti-CD99L2 Rabbit Polyclonal Antibody (Cy5®)

Dodavatel: Bioss
BS-13742R-CY5
BOSSBS-13742R-CY5EA 25630 CZK
BOSSBS-13742R-CY5
Anti-CD99L2 Rabbit Polyclonal Antibody (Cy5®)
Protilátky
Plays a role in a late step of leukocyte extravasation helping cells to overcome the endothelial basement membrane. Acts at the same site as, but independently of, PECAM1 (By similarity). Homophilic adhesion molecule, but these interactions may not be required for cell aggregation (By similarity).

Type: Primary
Antigen: CD99L2
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 81-187/262
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Produktová specifikace

Název antigenu CD99L2
Environmentally Preferable
Synonyma antigenu CD99B|CD99|UNQ1964/PRO4486|CD99L2|CD99 antigen-like protein 2|MIC2-like protein 1|MIC2L1
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Cy5®
Reaktivita Human
Hostitel Rabbit
Identifikační číslo genu 83692
Izotyp IgG
Western blot Yes
Imunochemie Yes
ImunoFluorescence Yes
Modifikace Unmodified
Epitop 81-187/262
Imunogen KLH conjugated synthetic peptide derived from human CD99L2
Formulář Liquid
Molekulární hmotnost 25 kDa
Čištění Purified by Protein A
Koncentrace 1 µg/µl
Skladovací pufr Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
Skladovací teplota Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Transportní teplota 4 °C
Testované aplikace ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR