Tisk…

Anti-MS4A6A Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Dodavatel: Bioss
BS-13692R-A750
BOSSBS-13692R-A750EA 25340 CZK
BOSSBS-13692R-A750
Anti-MS4A6A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Protilátky
MS4A6A is a member of the membrane-spanning 4A gene family and may be involved in signal transduction as a component of a multimeric receptor complex. There are three named isoforms.

Type: Primary
Antigen: MS4A6A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/248
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR