Tisk…

Anti-MS4A6A Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Dodavatel: Bioss
BS-13692R-A555
BOSSBS-13692R-A555EA 25430 CZK
BOSSBS-13692R-A555
Anti-MS4A6A Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Protilátky
MS4A6A is a member of the membrane-spanning 4A gene family and may be involved in signal transduction as a component of a multimeric receptor complex. There are three named isoforms.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MS4A6A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR