Tisk…

Anti-ZAK/HCCS4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Dodavatel: Bioss
BS-13546R-A750
BOSSBS-13546R-A750EA 25340 CZK
BOSSBS-13546R-A750
Anti-ZAK/HCCS4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Protilátky
Stress-activated component of a protein kinase signal transduction cascade. Regulates the JNK and p38 pathways. Pro-apoptotic. Role in regulation of S and G2 cell cycle checkpoint by direct phosphorylation of CHEK2. Isoform 1, but not isoform 2, causes cell shrinkage and disruption of actin stress fibers. Isoform 1 may have role in neoplastic cell transformation and cancer development. Isoform 1, but not isoform 2, phosphorylates histone H3 at 'Ser-28'.

Type: Primary
Antigen: ZAK/HCCS4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/800
Host: Rabbit
More Product Information
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR