Tisk…

Anti-GZMB Rabbit Polyclonal Antibody (Cy7®)

Dodavatel: Bioss
BS-1351R-CY7
BOSSBS-1351R-CY7EA 25890 CZK
BOSSBS-1351R-CY7
Anti-GZMB Rabbit Polyclonal Antibody (Cy7®)
Protilátky
This enzyme is necessary for target cell lysis in cell-mediated immune responses. It cleaves after Asp. Seems to be linked to an activation cascade of caspases (aspartate-specific cysteine proteases) responsible for apoptosis execution. Cleaves caspase-3, -7, -9 and 10 to give rise to active enzymes mediating apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: pan Granzyme
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR