Tisk…
Anti-GOLGA4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Dodavatel: Bioss
BS-13487R-FITC
BOSSBS-13487R-FITCEA 25630 CZK
BOSSBS-13487R-FITC
Anti-GOLGA4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Golgin 245 is also known as p230, GCP2, GOLG or golgi autoantigen and is a 2,230 amino acid protein that is expressed as three isoforms. Golgin 245 is localized to the cytoplasm in cells and is a member of the golgin family, all of which are peripheral membrane proteins associated with the Golgi complex. Golgin 245 has a carboxyl-terminal GRIP domain, which attaches to the trans-Golgi network (TGN) and TGN-derived vesicles. It is thought that the interaction that takes place between golgin 245 and MACF1, which cross-links microtubules to the Actin cytoskeleton, allows proteins to beMore Product Information