Tisk…

Anti-TRP73 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Dodavatel: Bioss
BS-1346R-A647
BOSSBS-1346R-A647EA 25660 CZK
BOSSBS-1346R-A647
Anti-TRP73 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
Participates in the apoptotic response to DNA damage. Isoforms containing the transactivation domain are pro-apoptotic, isoforms lacking the domain are anti-apoptotic and block the function of p53 and transactivating p73 isoforms. May be a tumor suppressor protein.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TRP73
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR