Tisk…
Anti-GOX Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Dodavatel: Bioss
BS-13387R-A555
BOSSBS-13387R-A555EA 25430 CZK
BOSSBS-13387R-A555
Anti-GOX Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Protilátky
Glucose Oxidase is a dimeric enzyme that binds to β-D-glucose and aids in its oxidation into D-glucono-1,5-lactone which then hydrolyzes to gluconic acid. Flavin adenine dinucleotide (FAD) is a cofactor to Glucose Oxidase that acts as the initial electron acceptor and is required for this oxidation to occur. Glucose Oxidase is a natural preservative found in honey, where it reduces atmospheric oxygen into hydrogen peroxide which acts as an antibacterial barrier. Glucose Oxidase is also commonly used in biosensors in which it conveys levels of glucose by keeping track of the number ofMore Product Information