Tisk…
Anti-GLDC Rabbit Polyclonal Antibody (Cy3®)
Dodavatel: Bioss
BS-13370R-CY3
BOSSBS-13370R-CY3EA 25630 CZK
BOSSBS-13370R-CY3
Anti-GLDC Rabbit Polyclonal Antibody (Cy3®)
Protilátky
The glycine cleavage system is comprised of AMT (known as Protein T), GCSH (known as Protein H), DLD (known as Protein L) and GLDC (known as Protein P), all of which work together to catalyze the cleavage and degradation of glycine. GLDC (glycine dehydrogenase ), also known as GCE, GCSP (glycine cleavage system P protein) or HYGN1, is a 1,020 amino acid protein that localizes to the mitochondria and belongs to the gcvP family. GLDC binds to glycine and enables the methylamine group from glycine to be transferred to the Protein T. GLDC exists as a homodimer and utilizes pyridoxal phosphate asMore Product Information