Tisk…
Anti-GEMIN6 Rabbit Polyclonal Antibody (Cy5®)
Dodavatel: Bioss
BS-13333R-CY5
BOSSBS-13333R-CY5EA 25630 CZK
BOSSBS-13333R-CY5
Anti-GEMIN6 Rabbit Polyclonal Antibody (Cy5®)
Protilátky
Spinal muscular atrophy (SMA) is an autosomal recessive neurodegenerative disease characterized by loss of motor neurons in the spinal cord. SMA is caused by deletion or loss-of-function mutations in the SMN (Survival of Motor Neuron) gene. Gemin6, the protein product of human chromosome 2p22.2, associates directly with SMN and is a part of the SMN complex containing Gemin2, Gemin3, Gemin4 and Gemin5 as well as several spliceosomal snRNP proteins. The SMN complex plays an essential role in spliceosomal snRNP assembly in the cytoplasm and is required for pre-mRNA splicing of the nucleus. TheMore Product Information