Tisk…
Anti-Gemin 6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Dodavatel: Bioss
BS-13333R-A680
BOSSBS-13333R-A680EA 25340 CZK
BOSSBS-13333R-A680
Anti-Gemin 6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Protilátky
Spinal muscular atrophy (SMA) is an autosomal recessive neurodegenerative disease characterized by loss of motor neurons in the spinal cord. SMA is caused by deletion or loss-of-function mutations in the SMN (Survival of Motor Neuron) gene. Gemin6, the protein product of human chromosome 2p22.2, associates directly with SMN and is a part of the SMN complex containing Gemin2, Gemin3, Gemin4 and Gemin5 as well as several spliceosomal snRNP proteins. The SMN complex plays an essential role in spliceosomal snRNP assembly in the cytoplasm and is required for pre-mRNA splicing of the nucleus. TheMore Product Information