Tisk…
Anti-GEMIN7 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Dodavatel: Bioss
BS-13330R-A350
BOSSBS-13330R-A350EA 25430 CZK
BOSSBS-13330R-A350
Anti-GEMIN7 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
Gemin7 is a 131 amino acid protein encoded by the mouse gene Gemin7. Gemin7, along with Gemin6, is a significant component of the the large multiprotein human SMN complex. The SMN complex functions as an assembly machine for small nuclear ribonucleoproteins (snRNPs)-the major components of the spliceosome. The survival of motor neurons (SMN) protein, a product of the disease gene of the common neurodegenerative disease, spinal muscular atrophy, is also part of the SMN complex. Although Gemin6 and Gemin7 have no significant sequence similarity with Sm proteins, both adopt canonical Sm folds.More Product Information