Tisk…
Anti-GALK2/Galactokinase 2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Dodavatel: Bioss
BS-13267R-A680
BOSSBS-13267R-A680EA 25340 CZK
BOSSBS-13267R-A680
Anti-GALK2/Galactokinase 2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Protilátky
GALK2 is important in the first step of the galactose metabolism pathway. GALK1, which belongs to the GHMP kinase family of proteins, is a crucial enzyme for galactose metabolism, specifically converting ?d-galactose to galactose 1-phosphate. Defects in the gene encoding GALK1 can cause galactosemia II, an autosomal recessive disorder characterized by congenital cataracts during infancy, often within the first two weeks of life. In the adult population, galactosemia II can cause presenile cataracts that are secondary to accumulation of galactitol in the lens of the eye. A second gene, GALK2,More Product Information