Tisk…

Anti-FRG1 Rabbit Polyclonal Antibody (Cy5®)

Dodavatel: Bioss
BS-13215R-CY5
BOSSBS-13215R-CY5EA 25630 CZK
BOSSBS-13215R-CY5
Anti-FRG1 Rabbit Polyclonal Antibody (Cy5®)
Protilátky
This gene maps to a location 100 kb centromeric of the repeat units on chromosome 4q35 which are deleted in facioscapulohumeral muscular dystrophy (FSHD). It is evolutionarily conserved and has related sequences on multiple human chromosomes but DNA sequence analysis did not reveal any homology to known genes. In vivo studies demonstrate the encoded protein is localized to the nucleolus. [provided by RefSeq, Jul 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FRG1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR