Tisk…
Anti-FCGBP Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Dodavatel: Bioss
BS-13168R-A350
BOSSBS-13168R-A350EA 25430 CZK
BOSSBS-13168R-A350
Anti-FCGBP Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
Fc (Ig constant fragment) receptors ensure protection of the host against foreign antigens, such as microorganisms and pathogens, by removing Ig-coated antigen complexes from circulation. Fc receptors are present on lymphoid and myeloid derivatives, where they mediate endocytosis of Ig-antigen complexes, antibody production in B cells through T cell antigen presentation, cytotoxicity and the release of cytokines and reactive oxygen species. The Fc γ-binding protein (FCGBP) interacts with the Fc portion of IgG and MUC2 to mediate the maintenance of the mucosal structure. FCGBP is a 5,405More Product Information