Tisk…
Anti-FBLN7 Rabbit Polyclonal Antibody (Cy7®)
Dodavatel: Bioss
BS-13161R-CY7
BOSSBS-13161R-CY7EA 25630 CZK
BOSSBS-13161R-CY7
Anti-FBLN7 Rabbit Polyclonal Antibody (Cy7®)
Protilátky
Fibulin-7 is a 439 amino acid extracellular matrix protein that belongs to the Fibulin family. Containing two EGF-like domains and one sushi (CCP/SCR) domain, Fibulin-7 exists as four alternatively spliced isoforms. Fibulin-7 is considered an adhesion protein that interacts with extracellular matrix molecules in developing teeth, and may be involved in differentiation and maintenance of odontoblasts as well as in dentin formation. Fibulin-7 is post-translationally glycosylated with N-linked oligosaccharides and interacts with heparin, fibronectin, fibulin-1 and DSP (dentinMore Product Information