Tisk…
Anti-ERF Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Dodavatel: Bioss
BS-13096R-FITC
BOSSBS-13096R-FITCEA 25630 CZK
BOSSBS-13096R-FITC
Anti-ERF Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
ABT1 (activator of basal transcription 1) is a nuclear protein that associates with the TATA-binding protein (TBP) and enhances basal transcription activity of class II promoters. ABT1 associates with TBP in HeLa nuclear extracts in vitro. Another protein, designated ERF, is a member of the Ets family of transcription factors. The members of the Ets family are grouped because they share a highly conserved DNA binding domain. These factors are involved in growth factor pathways and regulate both proliferation and differentiation. ERF (Ets-2 repressor factor) is a ubiquitously expressed Ets-More Product Information