Tisk…
Anti-SLC1A6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Dodavatel: Bioss
BS-13046R-HRP
BOSSBS-13046R-HRPEA 25630 CZK
BOSSBS-13046R-HRP
Anti-SLC1A6 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Excitatory Amino Acid Transporters (EAATs) are membrane-bound proteins that are localized in glial cells and pre-synaptic glutamatergic nerve endings. EAATs transport the excitatory neurotransmitters L-glutamate and D-aspartate, a process that is essential for terminating the postsynaptic action of glutamate. The re-uptake of amino acid neurotransmitters by EAAT proteins has been shown to protect neurons from excitotoxicity, which is caused by the accumulation of amino acid neurotransmitters. EAAT4 is an aspartate/glutamate transporter that is expressed predominantly in the cerebellum. TheMore Product Information