Tisk…

Anti-DIAPH2 Rabbit Polyclonal Antibody (Cy3®)

Dodavatel: Bioss
BS-13002R-CY3
BOSSBS-13002R-CY3EA 25630 CZK
BOSSBS-13002R-CY3
Anti-DIAPH2 Rabbit Polyclonal Antibody (Cy3®)
Protilátky
Could be involved in oogenesis. Involved in the regulation of endosome dynamics. Implicated in a novel signal transduction pathway, in which isoform 3 and CSK are sequentially activated by RHOD to regulate the motility of early endosomes through interactions with the actin cytoskeleton.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DIAPH2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR