Tisk…
Anti-KLHL28 Rabbit Polyclonal Antibody
Dodavatel: Bioss
BS-12907R
BOSSBS-12907REA 20650 CZK
BOSSBS-12907R
Anti-KLHL28 Rabbit Polyclonal Antibody
Protilátky
BTBD14A is a 587 amino acid protein that contains one BTB/POZ domain. The BTB/POZ domain mediates homomeric and heteromeric POZ-POZ interactions and is common to transcriptional regulators involved in chromatin modeling. In several BTB/POZ containing proteins, including BCL-6 and the promyelocytic leukemia zinc-finger (PLZF) oncoprotein, this domain interacts with the SMRT/N-CoR-mSin3A HDAC complex and is directly involved in repressing and silencing gene transcription. When this domain is deleted, as with the oncogenic PLZF-RAR chimera of promyelocytic leukemias, this transcriptionalMore Product Information