Tisk…
Anti-POU2AF1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Dodavatel: Bioss
BS-12876R-A350
BOSSBS-12876R-A350EA 25430 CZK
BOSSBS-12876R-A350
Anti-POU2AF1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
POU domain proteins contain a bipartite DNA-binding domain divided by a flexible linker that enables them to adopt various monomer configurations on DNA. The versatility of POU protein operation is additionally conferred at the dimerization level. The POU dimer from the OCT1 gene formed on the palindromic OCT factor recognition element, or PORE (ATTTGAAATGCAAAT), could recruit the transcriptional coactivator OBF1. Studies of tissue-specific expression of immunoglobulin promoters demonstrate the importance of an octamer, ATTTGCAT, and the proteins that bind to it. This is a regulatory elementMore Product Information