Tisk…
Anti-AQP3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Dodavatel: Bioss
BS-1253R-A350
BOSSBS-1253R-A350EA 25660 CZK
BOSSBS-1253R-A350
Anti-AQP3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
Water channel required to promote glycerol permeability and water transport across cell membranes. Acts as a glycerol transporter in skin and plays an important role in regulating SC (stratum corneum) and epidermal glycerol content. Involved in skin hydration, wound healing, and tumorigenesis. Provides kidney medullary collecting duct with high permeability to water, thereby permitting water to move in the direction of an osmotic gradient. Slightly permeable to urea and may function as a water and urea exit mechanism in antidiuresis in collecting duct cells. It may play an important role inMore Product Information